(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Leverandør: Apollo Scientific

OR32655-1G
APOSOR32655-1GEA 19275 NOK
APOSOR32655-1G
(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₃H₁₄N₂O
MW: 214,263 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01815113
CAS-nummer: 273748-55-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR