4-{[(1E)-2-Cyanoeth-1-en-1-yl]amino}benzonitrile, tech

Leverandør: Apollo Scientific

OR32864-1G
APOSOR32864-1GEA 19275 NOK
APOSOR32864-1G
4-{[(1E)-2-Cyanoeth-1-en-1-yl]amino}benzonitrile, tech
4-{[(1E)-2-Cyanoeth-1-en-1-yl]amino}benzonitrile
Formel: C₁₀H₇N₃
MW: 169,183 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00663726
CAS-nummer: 338959-90-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR