1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol, tech

Leverandør: Apollo Scientific

OR32915-1G
APOSOR32915-1GEA 19275 NOK
APOSOR32915-1G
1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol, tech
1-[4-(1H-Pyrrol-1-yl)phenyl]ethan-1-ol
Formel: C₁₂H₁₃NO
MW: 187,238 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03012945
CAS-nummer: 439110-83-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR