2-(1H-Pyrrol-1-yl)pyrazine 95%

Leverandør: Apollo Scientific

OR33001-1G
APOSOR33001-1GEA 10750 NOK
APOSOR33001-1G
2-(1H-Pyrrol-1-yl)pyrazine 95%
2-(1H-Pyrrol-1-yl)pyrazine
Formel: C₈H₇N₃
MW: 145,161 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01443653
CAS-nummer: 50966-75-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR