5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione, tech

Leverandør: Apollo Scientific

OR33040-1G
APOSOR33040-1GEA 19275 NOK
APOSOR33040-1G
5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione, tech
5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione
Formel: C₁₄H₁₆N₂O₂
MW: 244,289 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01310105
CAS-nummer: 419542-34-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR