(2Z)-2-[(4-Chlorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Leverandør: Apollo Scientific

OR33097-1G
APOSOR33097-1GEA 19275 NOK
APOSOR33097-1G
(2Z)-2-[(4-Chlorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(4-Chlorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₄H₁₄ClNO
MW: 247,72 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00664597
CAS-nummer: 54529-43-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR