(2E)-2-[(4-Methoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Leverandør: Apollo Scientific

OR33103-1G
APOSOR33103-1GEA 19275 NOK
APOSOR33103-1G
(2E)-2-[(4-Methoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2E)-2-[(4-Methoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₅H₁₇NO₂
MW: 243,301 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00665085
CAS-nummer: 52407-93-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR