2-[(3-Chlorophenyl)methoxy]ethan-1-ol, tech

Leverandør: Apollo Scientific

OR33269-1G
APOSOR33269-1GEA 19275 NOK
APOSOR33269-1G
2-[(3-Chlorophenyl)methoxy]ethan-1-ol, tech
2-[(3-Chlorophenyl)methoxy]ethan-1-ol
Formel: C₉H₁₁ClO₂
MW: 186,635 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02187353
CAS-nummer: 478257-99-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR