4-Amino-3,5-diphenyl-1,2,4-triazole 95%

Leverandør: Apollo Scientific

Synonymer: Diphenyl-4H-1,2,4-triazol-4-amine, 3,5-Diphenyl-4H-1,2,4-triazol-4-amine

OR33274-1G
APOSOR33274-1GEA 1375 NOK
APOSOR33274-1G
4-Amino-3,5-diphenyl-1,2,4-triazole 95%
4-Amino-3,5-diphenyl-1,2,4-triazole
Formel: C₁₄H₁₂N₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00450576
CAS-nummer: 3049-45-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR