3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile 95%

Leverandør: Apollo Scientific

OR33620-1G
APOSOR33620-1GEA 6625 NOK
APOSOR33620-1G
3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile 95%
3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile
Formel: C₁₀H₆BrN₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11100206
CAS-nummer: 1170378-39-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR