3-Chloro-5-nitropyridine-2-carboxylic acid 98%

Leverandør: Apollo Scientific

Synonymer: 3-Chloro-5-nitropicolinic acid

OR345612-250MG
APOSOR345612-250MGEA 1900 NOK
APOSOR345612-250MG
3-Chloro-5-nitropyridine-2-carboxylic acid 98%
3-Chloro-5-nitropyridine-2-carboxylic acid
Formel: C₆H₃ClN₂O₄
MW: 202,552 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD14584667
CAS-nummer: 141238-23-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR