2-(Chloromethyl)benzoyl chloride

Leverandør: Apollo Scientific
Danger

OR346332-5G
APOSOR346332-5GEA 950 NOK
APOSOR346332-5G
2-(Chloromethyl)benzoyl chloride
2-(Chloromethyl)benzoyl chloride
Formel: C₈H₆Cl₂O
MW: 189.04 g/mol
Smeltepunkt: 135 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00152348
CAS-nummer: 42908-86-1
EINECS: 255-998-2
FN: 2927
ADR: 6.1+8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR