5-Bromo-nicotinamidine hydrochloride

Leverandør: Apollo Scientific

OR346352-1G
APOSOR346352-1GEA 2500 NOK
APOSOR346352-1G
5-Bromo-nicotinamidine hydrochloride
5-Bromo-nicotinamidine hydrochloride
Formel: C₆H₆BrN₃·ClH
MW: 236,497 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD24386136
CAS-nummer: 1431555-24-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR