3-Chloro-o-toluidine

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Amino-6-chlorotoluene, 3-Chloro-2-methylaniline

OR350077-500G
APOSOR350077-500GEA 715 NOK
APOSOR350077-500G
3-Chloro-o-toluidine
3-Chloro-o-toluidine
Formel: C₇H₈ClN
Kokepunkt: 115…117 °C
Tetthet: 1,185 g/cm³ (25 °C)
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007766
CAS-nummer: 87-60-5
FN: 3429
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR