Diphenylphosphine oxide 97%

Leverandør: Apollo Scientific

OR350120-100G OR350120-25G OR350120-500G
APOSOR350120-100GEA 1550 NOK
APOSOR350120-100G APOSOR350120-25G APOSOR350120-500G
Diphenylphosphine oxide 97%
Diphenylphosphine oxide
Formel: C₁₂H₁₁OP
Kokepunkt: 105 °C/0,1mmHg
Smeltepunkt: 56…57 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002079
CAS-nummer: 4559-70-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR