2,5-Dibromophenylboronic acid

Leverandør: Apollo Scientific

OR350147-5G
APOSOR350147-5GEA 930 NOK
APOSOR350147-5G
2,5-Dibromophenylboronic acid
2,5-Dibromophenylboronic acid
Formel: C₆H₅BBr₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09743927
CAS-nummer: 1008106-93-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR