(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 95%

Leverandør: Apollo Scientific

OR350342-25G
APOSOR350342-25GEA 1100 NOK
APOSOR350342-25G
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 95%
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Formel: C₁₄H₁₅NO
MW: 213.28 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00074959
CAS-nummer: 23364-44-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR