Sodium-2-naphthol-7-sulphonate hydrate 98%

Leverandør: Apollo Scientific

OR350346-100G OR350346-500G
APOSOR350346-100GEA 805 NOK
APOSOR350346-100G APOSOR350346-500G
Sodium-2-naphthol-7-sulphonate hydrate 98%
Sodium-2-naphthol-7-sulphonate hydrate
Formel: C₁₀H₈O₄S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00021625
CAS-nummer: 135-55-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR