N-Methylpyrrole-3-boronic acid pinacol ester 97%

Leverandør: Apollo Scientific

Synonymer: 1-METHYL-1H-PYRROL-3-YLBORONIC ACID PINACOL ESTER, 1H-Pyrrole,1-methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-, 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

OR360175-1G
APOSOR360175-1GEA 540 NOK
APOSOR360175-1G
N-Methylpyrrole-3-boronic acid pinacol ester 97%
N-Methylpyrrole-3-boronic acid pinacol ester
Formel: C₁₁H₁₈BNO₂
MW: 207.08 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD13182200
CAS-nummer: 953040-54-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR