3-(1-Pyrrolidinylcarbonyl)phenylboronic acid 97%

Leverandør: Apollo Scientific
Warning

OR3976-5G
APOSOR3976-5GEA 2400 NOK
APOSOR3976-5G
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid 97%
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid
Formel: C₁₁H₁₄BNO₃
Smeltepunkt: 113…116 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04115685
CAS-nummer: 723281-53-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR