3-Amino-4-butoxybenzoic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Butoxy-5-carboxyaniline

OR40322-1G OR40322-250MG OR40322-500MG
APOSOR40322-1GEA 3775 NOK
APOSOR40322-1G APOSOR40322-250MG APOSOR40322-500MG
3-Amino-4-butoxybenzoic acid
3-Amino-4-butoxybenzoic acid
Formel: C₁₁H₁₅NO₃
MW: 209.242 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD12168705
CAS-nummer: 855202-63-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR