Orotic acid monohydrate

Leverandør: Apollo Scientific
Warning

Synonymer: Orotic acid hydrate, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate

OR40619-100G
APOSOR40619-100GEA 270 NOK
APOSOR40619-100G
Orotic acid monohydrate
Orotic acid monohydrate
Formel: C₅H₄N₂O₄·1H₂O
MW: 156,10 g/mol (anhy)
Smeltepunkt: 334 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149398
CAS-nummer: 50887-69-9
EINECS: 200-619-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR