trans-4-Hydroxy-D-proline 97%

Leverandør: Apollo Scientific
Warning

Synonymer: (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, H-D-Hyp-OH, H-trans-D-Hyp-OH

OR40650-5G
APOSOR40650-5GEA 2150 NOK
APOSOR40650-5G
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline
Formel: C₅H₉NO₃
Smeltepunkt: 273 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00065608
CAS-nummer: 3398-22-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR