1,3-Bis(difenylfosfino)propan 97%

Leverandør: Apollo Scientific
Warning

Synonymer: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

OR41032-5G OR41032-25G OR41032-100G
APOSOR41032-5GEA 270 NOK
APOSOR41032-5G APOSOR41032-25G APOSOR41032-100G
1,3-Bis(difenylfosfino)propan 97%
1,3-Bis(difenylfosfino)propan
Formel: C₂₇H₂₆P₂
MW: 412.45 g/mol
Smeltepunkt: 63…65 °C
MDL nummer: MFCD00003050
CAS-nummer: 6737-42-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR