2-(Furfurylthio)ethylamine

Leverandør: Apollo Scientific
Warning

OR42233-1G OR42233-250MG OR42233-500MG OR42233-5G
APOSOR42233-1GEA 645 NOK
APOSOR42233-1G APOSOR42233-250MG APOSOR42233-500MG APOSOR42233-5G
2-(Furfurylthio)ethylamine
2-(Furfurylthio)ethylamine
Formel: C₇H₁₁NOS
Tetthet: 1,13 g/cm³ (25 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01075029
CAS-nummer: 36415-21-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR