cis-Aconitic acid 90%

Leverandør: Apollo Scientific
Warning

Synonymer: (Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (1Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid

OR4293-1G OR4293-250MG
APOSOR4293-1GEA 1600 NOK
APOSOR4293-1G APOSOR4293-250MG
cis-Aconitic acid 90%
cis-Aconitic acid
Formel: C₆H₆O₆
MW: 174.11 g/mol
Smeltepunkt: 115…122 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00063184
CAS-nummer: 585-84-2
EINECS: 209-561-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR