(R)-(4-Benzylmorpholin-3-yl)methanol

Leverandør: Apollo Scientific

OR451054-1G
APOSOR451054-1GEA 1550 NOK
APOSOR451054-1G
(R)-(4-Benzylmorpholin-3-yl)methanol
(R)-(4-Benzylmorpholin-3-yl)methanol
Formel: C₁₂H₁₇NO₂
MW: 207,2700 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06799481
CAS-nummer: 101376-26-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR