(S)-(4-Benzylmorpholin-3-yl)methanol

Leverandør: Apollo Scientific

OR451055-1G
APOSOR451055-1GEA 875 NOK
APOSOR451055-1G
(S)-(4-Benzylmorpholin-3-yl)methanol
(S)-(4-Benzylmorpholin-3-yl)methanol
Formel: C₁₂H₁₇NO₂
MW: 207,269 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06799482
CAS-nummer: 101376-25-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR