(S)-4-Boc-3-(aminomethyl)-morpholine

Leverandør: Apollo Scientific

Synonymer: (S)-tert-Butyl 3-(aminomethyl)morpholine-4-carboxylate

OR451063-1G
APOSOR451063-1GEA 18850 NOK
APOSOR451063-1G
(S)-4-Boc-3-(aminomethyl)-morpholine
(S)-4-Boc-3-(aminomethyl)-morpholine
Formel: C₁₀H₂₀N₂O₃
MW: 216,277 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06796250
CAS-nummer: 1187929-79-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR