(2S)-2-Amino-2-cyclobutylethan-1-ol hydrochloride

Leverandør: Apollo Scientific

OR451078-500MG
APOSOR451078-500MGEA 16150 NOK
APOSOR451078-500MG
(2S)-2-Amino-2-cyclobutylethan-1-ol hydrochloride
(2S)-2-Amino-2-cyclobutylethan-1-ol hydrochloride
Formel: C₆H₁₃NO·ClH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28975217
CAS-nummer: 1259702-72-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR