(S)-2-Benzylpiperazine dihydrochloride

Leverandør: Apollo Scientific

OR451101-1G
APOSOR451101-1GEA 5775 NOK
APOSOR451101-1G
(S)-2-Benzylpiperazine dihydrochloride
(S)-2-Benzylpiperazine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
MW: 249,18 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09954328
CAS-nummer: 208655-19-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR