(S)-2-Cyclobutylglycine

Leverandør: Apollo Scientific

Synonymer: (S)-2-Amino-2-cyclobutylacetic acid

OR451131-1G
APOSOR451131-1GEA 3575 NOK
APOSOR451131-1G
(S)-2-Cyclobutylglycine
(S)-2-Cyclobutylglycine
Formel: C₆H₁₁NO₂
MW: 129,157 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08436202
CAS-nummer: 49607-08-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR