6-Amino-3,4-dihydroquinolin-2(1H)-one

Leverandør: Apollo Scientific

OR451145-5G
APOSOR451145-5GEA 7250 NOK
APOSOR451145-5G
6-Amino-3,4-dihydroquinolin-2(1H)-one
6-Amino-3,4-dihydroquinolin-2(1H)-one
Formel: C₉H₁₀N₂O
MW: 162,189 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00559318
CAS-nummer: 22246-13-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR