1-Boc-4-ethynyl-1H-pyrazole

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Methyl-2-propanyl 4-ethynyl-1H-pyrazole-1-carboxylate

OR451236-250MG
APOSOR451236-250MGEA 3575 NOK
APOSOR451236-250MG
1-Boc-4-ethynyl-1H-pyrazole
1-Boc-4-ethynyl-1H-pyrazole
Formel: C₁₀H₁₂N₂O₂
MW: 192,214 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD16293741
CAS-nummer: 1093193-29-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR