(S)-3-Benzylmorpholine hydrochloride

Leverandør: Apollo Scientific

OR451243-1G
APOSOR451243-1GEA 3100 NOK
APOSOR451243-1G
(S)-3-Benzylmorpholine hydrochloride
(S)-3-Benzylmorpholine hydrochloride
Formel: C₁₁H₁₅NO·ClH
MW: 213,704 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18381984
CAS-nummer: 77897-23-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR