4-Chloro-1-methyl-1H-pyrazol-3-amine hydrochloride

Leverandør: Apollo Scientific
Warning

OR451441-1G OR451441-5G
APOSOR451441-1GEA 790 NOK
APOSOR451441-1G APOSOR451441-5G
4-Chloro-1-methyl-1H-pyrazol-3-amine hydrochloride
4-Chloro-1-methyl-1H-pyrazol-3-amine hydrochloride
Formel: C₄H₆ClN₃·ClH
MW: 168.024 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD15142897
CAS-nummer: 1197235-09-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR