(R)-(+)-3-Hydroxypiperidine hydrochloride 98%

Leverandør: Apollo Scientific

Synonymer: (R)-3-Piperidinol hydrochloride, (3R)-piperidin-3-ol hydrochloride, (R)-Piperidin-3-ol hydrochloride, (R)-3-Hydroxypiperidine hydrochloride

OR4578-25G
APOSOR4578-25GEA 1375 NOK
APOSOR4578-25G
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
(R)-(+)-3-Hydroxypiperidine hydrochloride
Formel: C₅H₁₂ClNO
MW: 137,607 g/mol
Smeltepunkt: 195…196 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00192221
CAS-nummer: 198976-43-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR