3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 2-(3-Oxo-2H-benzo[b][1,4]oxazin-4(3H)-yl)acetic acid

OR4593-250MG
APOSOR4593-250MGEA 450 NOK
APOSOR4593-250MG
3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
Formel: C₁₀H₉NO₄
MW: 207,19 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00269587
CAS-nummer: 26494-55-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR