cis-1,3-Cyclopentanedicarboxylic acid

Leverandør: Apollo Scientific
Warning

OR46133-1G OR46133-5G
APOSOR46133-1GEA 380 NOK
APOSOR46133-1G APOSOR46133-5G
cis-1,3-Cyclopentanedicarboxylic acid
cis-1,3-Cyclopentanedicarboxylic acid
Formel: C₇H₁₀O₄
Smeltepunkt: 121 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01311175
CAS-nummer: 876-05-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR