3-Picolyl bromide hydrobromide 96%

Leverandør: Apollo Scientific
Danger

OR46547-25G
APOSOR46547-25GEA 1325 NOK
APOSOR46547-25G
3-Picolyl bromide hydrobromide 96%
3-Picolyl bromide hydrobromide
Formel: C₆H₇Br₂N
Smeltepunkt: 150…155 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01321314
CAS-nummer: 4916-55-6
FN: 3261
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR