3-Dimethylamino-2,2-dimethylpropionaldehyde

Leverandør: Apollo Scientific

OR46639-250MG
APOSOR46639-250MGEA 2100 NOK
APOSOR46639-250MG
3-Dimethylamino-2,2-dimethylpropionaldehyde
3-Dimethylamino-2,2-dimethylpropionaldehyde
Formel: C₇H₁₅NO
MW: 129.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00043600
CAS-nummer: 15451-14-6
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR