2-Hydroxybenzoyl chloride

Leverandør: Apollo Scientific
Danger

OR46678-1G OR46678-250MG OR46678-500MG
APOSOR46678-1GEA 7575 NOK
APOSOR46678-1G APOSOR46678-250MG APOSOR46678-500MG
2-Hydroxybenzoyl chloride
2-Hydroxybenzoyl chloride
Formel: C₇H₅ClO₂
MW: 156.57 g/mol
Smeltepunkt: 17…20 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11169645
CAS-nummer: 1441-87-8
FN: 1759
ADR: 8 III
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR