N-(2,5-Dimethyl-4-nitrophenyl)acetamide

Leverandør: Apollo Scientific

OR470041-5G
APOSOR470041-5GEA 5525 NOK
APOSOR470041-5G
N-(2,5-Dimethyl-4-nitrophenyl)acetamide
N-(2,5-Dimethyl-4-nitrophenyl)acetamide
Formel: C₁₀H₁₂N₂O₃
MW: 208.214 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00547733
CAS-nummer: 6954-69-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR