5-Amino-2-isopropyl-2H-indazole

Leverandør: Apollo Scientific

OR470057-5G
APOSOR470057-5GEA 13775 NOK
APOSOR470057-5G
5-Amino-2-isopropyl-2H-indazole
5-Amino-2-isopropyl-2H-indazole
Formel: C₁₀H₁₃N₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18375232
CAS-nummer: 1261268-91-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR