3-(tert-Butoxy)benzenecarboxylic acid

Leverandør: Apollo Scientific

Synonymer: 3-tert-Butoxybenzoic acid

OR470339-5G OR470339-1G
APOSOR470339-5GEA 13775 NOK
APOSOR470339-5G APOSOR470339-1G
3-(tert-Butoxy)benzenecarboxylic acid
3-(tert-Butoxy)benzenecarboxylic acid
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03412397
CAS-nummer: 15360-02-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR