Methyl 2-Amino-2-(4-biphenylyl)acetate

Leverandør: Apollo Scientific

OR470396-5G
APOSOR470396-5GEA 18850 NOK
APOSOR470396-5G
Methyl 2-Amino-2-(4-biphenylyl)acetate
Methyl 2-Amino-2-(4-biphenylyl)acetate
Formel: C₁₅H₁₅NO₂
MW: 241,285 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12518288
CAS-nummer: 515158-65-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR