Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate

Leverandør: Apollo Scientific

OR470457-5G
APOSOR470457-5GEA 6025 NOK
APOSOR470457-5G
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
Ethyl 1-Cbz-3-allylpiperidine-3-carboxylate
Formel: C₁₉H₂₅NO₄
MW: 331,406 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21641322
CAS-nummer: 1363166-44-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR