Methyl 1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylate

Leverandør: Apollo Scientific

OR470526-5G
APOSOR470526-5GEA 12400 NOK
APOSOR470526-5G
Methyl 1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₈O₄
MW: 262,301 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22423123
CAS-nummer: 63383-54-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR