Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate

Leverandør: Apollo Scientific

OR470529-5G
APOSOR470529-5GEA 12400 NOK
APOSOR470529-5G
Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₈O₄
MW: 262,301 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22423130
CAS-nummer: 1384264-82-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR