2-(Hydroxymethyl)-N-methylbenzamide

Leverandør: Apollo Scientific

OR470601-5G
APOSOR470601-5GEA 8775 NOK
APOSOR470601-5G
2-(Hydroxymethyl)-N-methylbenzamide
2-(Hydroxymethyl)-N-methylbenzamide
Formel: C₉H₁₁NO₂
MW: 165,189 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00157782
CAS-nummer: 39976-03-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR